• USA Home
  • Search Results

  • penteticacid393356743611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentetic acid

1 Products
Synonym:   (Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Diethylenetriaminepentaacetic acid, Penta(carboxymethyl)diethylenetriamine, Pentetic acid
Image of Pentetic acid
CAS Number:
Linear Formula:
 [(HOOCCH2)2NCH2CH2]2NCH2COOH
Molecular Weight:
 393.35
 Beilstein/REAXYS Number:
 1810219
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More