• USA Home
  • Search Results

  • pentobarbital226277674411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentobarbital

1 Products
Synonym:   5-Ethyl-5-(1-methylbutyl)-2,4,6-trioxohexahydropyrimidine, Nembutal
Image of Pentobarbital
CAS Number:
Empirical Formula (Hill Notation):
 C11H18N2O3
Molecular Weight:
 226.27
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More