• USA Home
  • Search Results

  • pentylacetate1301862863711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pentyl acetate

3 Products
Synonym:   Amyl acetate
Image of Pentyl acetate
CAS Number:
Linear Formula:
 CH3COO(CH2)4CH3
Molecular Weight:
 130.18
 Beilstein/REAXYS Number:
 1744753
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Application(s)
Show More