• USA Home
  • Search Results

  • peonidin3oglucosidechloride49886690639411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peonidin 3-O-glucoside chloride

2 Products
Synonym:   3-(Glucosyloxy)-4′,5,7-trihydroxy-3′-methoxyflavylium chloride, Glucopeonidin chloride
Image of Peonidin 3-<I>O</I>-glucoside chloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H23ClO11
Molecular Weight:
 498.86
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More