• USA Home
  • Search Results

  • peonidinchloride3367213401011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peonidin chloride

1 Products
Synonym:   3,4′,5,7-Tetrahydroxy-3′-methoxyflavylium chloride, Paeonidin chloride, Peonidol chloride
Image of Peonidin chloride
CAS Number:
Empirical Formula (Hill Notation):
 C16H13ClO6
Molecular Weight:
 336.72
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More