• USA Home
  • Search Results

  • peptonefromanimaltissue123457304973711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peptone from animal tissue

2 Products
CAS Number:
 MDL number:
 eCl@ss:
 42040102
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Suitability
  Product Line
Show More