• USA Home
  • Search Results

  • peptonefrommeatbacteriological123459107938811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peptone from meat, bacteriological

1 Products
Synonym:   Bacteriological peptone, Peptone from bovine meat
CAS Number:
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
Show More