• USA Home
  • Search Results

  • peptonefrommeatenzymaticdigest123459107938811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peptone from meat, enzymatic digest

1 Products
Synonym:   Peptone from bovine meat
CAS Number:
 eCl@ss:
 42040102
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Media Suitability
  Special Grade
  pH Value
  Solubility
  Loss
Show More