• USA Home
  • Search Results

  • peptonefromsoybeanenzymaticdigest123459107946811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peptone from soybean, enzymatic digest

3 Products
Synonym:   Peptone from soybean
CAS Number:
 EC Number:
 eCl@ss
 42040102
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Loss
  Special Grade
  pH Value
  Solubility
Show More