• USA Home
  • Search Results

  • peptonemycological123459107938811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peptone, mycological

1 Products
Synonym:   Mycological Peptone, Peptone from bovine meat
CAS Number:
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More