• USA Home
  • Search Results

  • perchloricacid10046760190311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perchloric acid

8 Products
Synonym:   PCA
Image of Perchloric acid
CAS Number:
Linear Formula:
 HClO4
Molecular Weight:
 100.46
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Quality clarifier
  Melting Point
  Ice Shipping
  Vapor pressure
  Purity Grade
  Color
  Restriction
  Agency/Method
  Concentration
  Physical Form
  Special Grade
  Density
Show More