• USA Home
  • Search Results

  • perfluorodecanoicacid5140833576211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perfluorodecanoic acid

2 Products
Synonym:   Nonadecafluorocapric acid, Nonadecafluorodecanoic acid, Perfluorocapric acid, Perfluorodecanoic acid
Image of Perfluorodecanoic acid
CAS Number:
Linear Formula:
 CF3(CF2)8CO2H
Molecular Weight:
 514.08
 Beilstein/REAXYS Number:
 1810811
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More