• USA Home
  • Search Results

  • perfluoropentanoicacid26405270690311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perfluoropentanoic acid

2 Products
Synonym:   Nonafluoropentanoic acid, Nonafluorovaleric acid, PFPeA, Perfluoropentanoic acid
Image of Perfluoropentanoic acid
CAS Number:
Linear Formula:
 CF3(CF2)3COOH
Molecular Weight:
 264.05
 Beilstein/REAXYS Number:
 1800087
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More