• USA Home
  • Search Results

  • perfluoropropyliodide2959375434711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perfluoropropyl iodide

1 Products
Synonym:   1-Iodoperfluoropropane, Heptafluoro-1-iodopropane, Heptafluoropropyl iodide
Image of Perfluoropropyl iodide
CAS Number:
Linear Formula:
 CF3CF2CF2I
Molecular Weight:
 295.93
 Beilstein/REAXYS Number:
 1758822
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More