• USA Home
  • Search Results

  • perfluoroundecanoicacid56409205894811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perfluoroundecanoic acid

2 Products
Synonym:   Heneicosafluoroundecanoic acid, PFUnDA
Image of Perfluoroundecanoic acid
CAS Number:
Linear Formula:
 CF3(CF2)9CO2H
Molecular Weight:
 564.09
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More