• USA Home
  • Search Results

  • peroxidasefromhorseradish12345900399011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Peroxidase from horseradish

9 Products
Synonym:   Detection Enzyme, Donor:hydrogen-peroxide oxidoreductase, HRP, Horseradish peroxidase, Peroxidase
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 1.11.1.7  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
 eCl@ss:
 32160410
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Ice Shipping
  Specific Activity
  Type
  Absorbance ratio
  Molecular weight
  Physical Form
Show More

Product #
Description
P8375 Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol),
P6782 Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol), 950-2000 units/mg solid (using ABTS),
P8250 Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol),
P8125 Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol),
77332 Peroxidase from horseradish, lyophilized, powder, beige, ~150 U/mg,
SRE0082 Peroxidase from horseradish, Suitable for manufacturing of diagnostic kits and reagents,
P2088 Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol),
P8415 Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol),
P6140 Peroxidase from horseradish, Type X, ammonium sulfate suspension,