• USA Home
  • Search Results

  • perylene2523119855011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Perylene

4 Products
Image of Perylene
CAS Number:
Linear Formula:
 C₂₀H₁₂
Molecular Weight:
 252.31 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Orbital energy
  Special Grade
  Titertest
  Fluorescence Test
Show More