• USA Home
  • Search Results

  • petrol12345803232411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Petrol

2 Products
Synonym:   Gasoline, Petroleum benzin
CAS Number:
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  Restriction
  Refractive Index
  Special Grade
  Assay
  Physical Form
  Purity Grade
Show More