• USA Home
  • Search Results

  • petroleumether1234510131646511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Petroleum ether

6 Products
Synonym:   Petroleum benzin
CAS Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  Miscellaneous Specifications
  Quality clarifier
  Special Grade
  Refractive Index
  Vapor pressure
  Vapor density vs air
  Purity Grade
Show More