• USA Home
  • Search Results

  • petroleumether12345803232411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Petroleum ether

5 Products
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.07
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Assay
  Color
  Vapor pressure
  UV-Absorption Values
  Product Line
  Physical Form
Show More