• USA Home
  • Search Results

  • phenanthrenesolution123458501811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenanthrene solution

1 Products
Image of Phenanthrene solution
CAS Number:
 Beilstein/REAXYS Number:
 1905428
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Purity Grade
  Package quantity
  Anal. Standard Form
  Special Grade
  Concentration
Show More