• USA Home
  • Search Results

  • phenelzinesulfatesalt2342715651411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenelzine sulfate salt

1 Products
Synonym:   Phenelzine hydrogen sulphate
Image of Phenelzine sulfate salt
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N2 · H2O4S
Molecular Weight:
 234.27
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P6777 Phenelzine sulfate salt,