• USA Home
  • Search Results

  • phenethylbenzoate226279447311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenethyl benzoate

2 Products
Image of Phenethyl benzoate
CAS Number:
Linear Formula:
 C6H5CO2CH2CH2C6H5
Molecular Weight:
 226.27
 Council of Europe no.:
 667
 EC Number:
 FEMA Number:
 2860
 Flavis number:
 9.774
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Biological Source
  Assay
  Special Grade
Show More