• USA Home
  • Search Results

  • phenethylisobutyrate1922510348011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenethyl isobutyrate

1 Products
Image of Phenethyl isobutyrate
CAS Number:
Linear Formula:
 (CH3)2CHCO2CH2CH2C6H5
Molecular Weight:
 192.25
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Special Grade
  Assay
Show More