• USA Home
  • Search Results

  • phenobarbitald5sidechainsolution237277373805311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenobarbital-D5 (side chain) solution

2 Products
Image of Phenobarbital-D<SUB>5 </SUB>(side chain) solution
CAS Number:
Empirical Formula (Hill Notation):
 C12D5H7N2O3
Molecular Weight:
 237.27
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More