• USA Home
  • Search Results

  • phenol941110895211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenol

19 Products
Synonym:   Hydroxybenzene, Carbolic acid, Hydroxybenzene
Image of Phenol
CAS Number:
Linear Formula:
 C6H5OH
Molecular Weight:
 94.11
 Beilstein/REAXYS Number:
 969616
 EC Index Number:
 203-632-7
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Anal. Standard Form
  Special Grade
  Allergen
  Organoleptic property
  Melting Point
  Media Suitability
  Application(s)
  Quality clarifier
  Purity Grade
  Potency
  Bulk Density
  Restriction
  Suitability
  Product Line
  pH Value
  Additive
  Transition temperature
  Solubility
  Agency/Method
  Physical Form
  Assay
Show More

Product #
Description
P1037 Phenol, for molecular biology,
328111 Phenol, unstabilized, purified by redistillation, ≥99%,
P5566 Phenol, BioXtra, ≥99.5% (GC),
242322 Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%,
185450 Phenol, unstabilized, ReagentPlus®, ≥99%,
77607 Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T),
W322318 Phenol, ≥99%,
33517 Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%,
77608 Phenol, BioUltra, for molecular biology, ≥99.5% (GC),
16017 Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC),
16016 Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached),
16018 Phenol, ≥96.0% (calc. on dry substance, T),
35952 Phenol, PESTANAL®, analytical standard,
1.00201 Phenol, EMPROVE® ESSENTIAL Ph Eur,ChP,JP,USP,
1.00206 Phenol, GR for analysis ACS,Reag. Ph Eur,
PHR1047 Phenol, Pharmaceutical Secondary Standard; Certified Reference Material,
1524806 Phenol, United States Pharmacopeia (USP) Reference Standard,
8.22296 Phenol, for synthesis,
W322340 Phenol, natural, 97%, FG,