• USA Home
  • Search Results

  • phenolphthaleinsolution318327709811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenolphthalein solution

1 Products
Image of Phenolphthalein solution
CAS Number:
Empirical Formula (Hill Notation):
 C20H14O4
Molecular Weight:
 318.32
 Beilstein/REAXYS Number:
 284423
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
  pH Value
Show More