• USA Home
  • Search Results

  • phenolsolution941110895211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenol solution

7 Products
Image of Phenol solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H6O
Molecular Weight:
 94.11
 Beilstein/REAXYS Number:
 6428463
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Refractive Index
  Purity Grade
  Package quantity
  Feature
  Density
  Anal. Standard Form
  Product Line
  Physical Form
  Special Grade
  Concentration
  Agency/Method
Show More