• USA Home
  • Search Results

  • phenothiazine199279284211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenothiazine

5 Products
Synonym:   10H-Phenothiazine
Image of Phenothiazine
CAS Number:
Empirical Formula (Hill Notation):
 C12H9NS
Molecular Weight:
 199.27
 Beilstein/REAXYS Number:
 143237
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
  Product Line
  Melting Point
  Special Grade
Show More