• USA Home
  • Search Results

  • phenterminehydrochloride18569119721311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phentermine hydrochloride

2 Products
Synonym:   α,α-Dimethylphenethylamine hydrochloride
Image of Phentermine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H15N · HCl
Molecular Weight:
 185.69
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
Show More