• USA Home
  • Search Results

  • phentolaminemethanesulfonatesalt377466528111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phentolamine methanesulfonate salt

1 Products
Synonym:   2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline
Image of Phentolamine methanesulfonate salt
CAS Number:
Linear Formula:
 C17H19N3O · CH3SO3H
Molecular Weight:
 377.46
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More