• USA Home
  • Search Results

  • phenylacetaldehyde1201512278111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylacetaldehyde

2 Products
Synonym:   α-Tolyaldehyde, 2-Phenylacetaldehyde
Image of Phenylacetaldehyde
CAS Number:
Linear Formula:
 C6H5CH2CHO
Molecular Weight:
 120.15
 Beilstein/REAXYS Number:
 385791
 Council of Europe no.:
 116
 EC Number:
 FEMA Number:
 2874
 Flavis number:
 5.030
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39023706
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Agency/Method
  Density
  Assay
  Special Grade
Show More