• USA Home
  • Search Results

  • phenylacetate1361512279211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenyl acetate

2 Products
Synonym:   Acetic acid phenyl ester
Image of Phenyl acetate
CAS Number:
Linear Formula:
 CH3COOC6H5
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 636458
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More