• USA Home
  • Search Results

  • phenylaceticacid1361510382211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylacetic acid

7 Products
Synonym:   Phenylacetic acid
Image of Phenylacetic acid
CAS Number:
Empirical Formula (Hill Notation):
 C8H8O2
Molecular Weight:
 136.15
 EC Index Number
 203-148-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
  Potency
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Special Grade
  Assay
Show More