• USA Home
  • Search Results

  • phenylacetylene1021353674311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylacetylene

2 Products
Synonym:   Ethynylbenzene
Image of Phenylacetylene
CAS Number:
Linear Formula:
 C6H5CCH
Molecular Weight:
 102.13
 Beilstein/REAXYS Number:
 605461
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More