• USA Home
  • Search Results

  • phenylephrinehydrochloride203676176711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylephrine hydrochloride

4 Products
Synonym:   (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride, (R)-(−)-Phenylephrine hydrochloride
Image of Phenylephrine hydrochloride
CAS Number:
Linear Formula:
 HOC6H4CH(CH2NHCH3)OH·HCl
Molecular Weight:
 203.67
 Beilstein/REAXYS Number:
 4158948
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More