• USA Home
  • Search Results

  • phenylglyoxalmonohydrate15215107412011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylglyoxal monohydrate

1 Products
Image of Phenylglyoxal monohydrate
CAS Number:
Linear Formula:
 C6H5COCHO · H2O
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 1854721
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More