• USA Home
  • Search Results

  • phenylphosphonicdichloride1949882472611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylphosphonic dichloride

1 Products
Synonym:   P,P-Dichlorophenylphosphine oxide, Benzenephosphonic dichloride, Benzenephosphonyl chloride, Phenyldichlorophosphine oxide, Phenylphosphonyl dichloride, Phenylphosphoryl dichloride
Image of Phenylphosphonic dichloride
CAS Number:
Linear Formula:
 C6H5POCl2
Molecular Weight:
 194.98
 Beilstein/REAXYS Number:
 1072240
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More