• USA Home
  • Search Results

  • phenylpyruvicacid1641615606911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phenylpyruvic acid

2 Products
Synonym:   2-Oxo-3-phenylpropanoic acid, 2-Oxo-3-phenylpropionic acid
Image of Phenylpyruvic acid
CAS Number:
Linear Formula:
 C6H5CH2COCOOH
Molecular Weight:
 164.16
 Beilstein/REAXYS Number:
 2207312
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More