• USA Home
  • Search Results

  • phloretin274276082211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phloretin

3 Products
Synonym:   β-(4-Hydroxyphenyl)-2,4,6-trihydroxypropiophenone, 2′,4′,6′-Trihydroxy-3-(4-hydroxyphenyl)propiophenone, 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)-1-propanone
Image of Phloretin
CAS Number:
Empirical Formula (Hill Notation):
 C15H14O5
Molecular Weight:
 274.27
 Beilstein/REAXYS Number:
 1887240
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More