• USA Home
  • Search Results

  • phloridzindihydrate47244706154311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phloridzin dihydrate

3 Products
Synonym:   1-[2-(β-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone, Phloretin 2′-β-D-glucopyranoside, Phloretin 2′-β-D-glucoside, Phlorizin dihydrate
Image of Phloridzin dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C21H24O10 · 2H2O
Molecular Weight:
 472.44
 Beilstein/REAXYS Number:
 66621
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More