• USA Home
  • Search Results

  • phloxineb829631847287211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phloxine B

2 Products
Synonym:   2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluorescein disodium salt, Acid Red 92, Cyanosine, D&C;Red No. 28, Eosin 10B
Image of Phloxine B
CAS Number:
Empirical Formula (Hill Notation):
 C20H2Br4Cl4Na2O5
Molecular Weight:
 829.63
 Beilstein/REAXYS Number:
 3887100
 Colour Index Number:
 45410
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More