• USA Home
  • Search Results

  • phorbol1213diacetate448512492815211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phorbol 12,13-diacetate

1 Products
Synonym:   PDA
Image of Phorbol 12,13-diacetate
CAS Number:
Empirical Formula (Hill Notation):
 C24H32O8
Molecular Weight:
 448.51
 Beilstein/REAXYS Number:
 2317312
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More