• USA Home
  • Search Results

  • phorbol12myristate13acetate616831656129811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phorbol 12-myristate 13-acetate

2 Products
Synonym:   12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA
Image of Phorbol 12-myristate 13-acetate
CAS Number:
Empirical Formula (Hill Notation):
 C36H56O8
Molecular Weight:
 616.83
 Beilstein/REAXYS Number:
 2407201
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More