• USA Home
  • Search Results

  • phosalone36781231017011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosalone

3 Products
Image of Phosalone
CAS Number:
Empirical Formula (Hill Notation):
 C12H15ClNO4PS2
Molecular Weight:
 367.81
 Beilstein/REAXYS Number:
 694650
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Package quantity
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More