• USA Home
  • Search Results

  • phosgenesolution98927544511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosgene solution

1 Products
Image of Phosgene solution
CAS Number:
Linear Formula:
 COCl2
Molecular Weight:
 98.92
 Beilstein/REAXYS Number:
 1098367
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More