• USA Home
  • Search Results

  • phosphamidon299691317121611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphamidon

2 Products
Synonym:   2-Chloro-N,N-diethyl-3-(dimethylphosphono)crotonic amide
Image of Phosphamidon
CAS Number:
Empirical Formula (Hill Notation):
 C10H19ClNO5P
Molecular Weight:
 299.69
 Beilstein/REAXYS Number:
 8323501
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Special Grade
Show More