• USA Home
  • Search Results

  • phosphataseacidfrompotato12345900177811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphatase, Acid from potato

3 Products
Synonym:   (Orthophosphoric-monoester phosphohydrolase acid optimum), Acid Phosphatase from potato
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.3.2  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
 eCl@ss:
 42010102
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Ice Shipping
  Molecular weight
Show More