• USA Home
  • Search Results

  • phosphataseacidfromwheatgerm12345900177811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphatase, Acid from wheat germ

1 Products
Synonym:   (Orthophosphoric-monoester phosphohydrolase [acid optimum] ), Acid Phosphatase from wheat germ
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.3.2  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
 eCl@ss:
 42010102
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More