• USA Home
  • Search Results

  • phosphocholinechloridecalciumsalttetrahydrate329737255674211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Phosphocholine chloride calcium salt tetrahydrate

1 Products
Synonym:   Calcium phosphorylcholine chloride, Choline chloride phosphate calcium salt, Choline phosphate chloride, Phosphorocholine chloride calcium salt, Phosphorylcholine chloride calcium salt
Image of Phosphocholine chloride calcium salt tetrahydrate
CAS Number:
Linear Formula:
 CaO3POCH2CH2N(Cl)(CH3)3 · 4H2O
Molecular Weight:
 329.73
 Beilstein/REAXYS Number:
 4835230
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More